Skip to Content
Merck
All Photos(1)

Key Documents

C2300000

Clofibrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(4-Chlorophenoxy)-2-methylpropionic acid ethyl ester, Ethyl 2-(4-chlorophenoxy)-2-methylpropionate, Ethyl 2-(4-chlorophenoxy)isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15ClO3
CAS Number:
Molecular Weight:
242.70
Beilstein:
1913459
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clofibrate

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1

InChI

1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3

InChI key

KNHUKKLJHYUCFP-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clofibrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review of studies on the mode of action of clofibrate and betabenzalbutyrate.
H J Fallon et al.
Lipids, 7(2), 106-109 (1972-02-01)
Clofibrate myopathy: a case report and a review of the literature.
P Rush et al.
Seminars in arthritis and rheumatism, 15(3), 226-229 (1986-02-01)
F Penna et al.
Current medicinal chemistry, 17(4), 309-320 (2009-12-18)
The biological activity of peroxisome proliferators (PPs) is mediated by a class of receptors, known as PPARs (PP-Activated Receptor), belonging to the nuclear receptor superfamily. Upon ligand binding, PPARs dimerize with retinoid receptors, translocate to the nucleus, recognize specific PP-responsive
H M Mehendale
Toxicological sciences : an official journal of the Society of Toxicology, 57(2), 187-190 (2000-09-28)
The article highlighted in this issue is "Peroxisome Proliferator-Activated Receptor Alpha-Null Mice Lack Resistance to Acetaminophen Hepatotoxicity Following Clofibrate Exposure" by Chuan Chen, Gayle E. Hennig, Herbert E. Whiteley, J Christopher Corton, and José E. Manautou (pp. 338-344).
S Oikawa et al.
Nihon rinsho. Japanese journal of clinical medicine, 52(12), 3285-3291 (1994-12-01)
Clofibrate has cholesterol- and triglyceride-lowering effect. They affect on the various points in the metabolic pathway of lipoproteins. They improve VLDL-synthesis in liver and increase the activity of LPL and hepatic TG lipase. As the results, HDL-cholesterol increases and LDL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service