Skip to Content
Merck
All Photos(2)

Key Documents

03422

Supelco

5-Tricosylresorcinol

analytical standard

Synonym(s):

1,3-Dihydroxy-5-tricosylbenzene, 5-Tricosyl-1,3-benzenediol, Tricosylresorcinol (23:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H52O2
CAS Number:
Molecular Weight:
432.72
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCc1cc(O)cc(O)c1

InChI

1S/C29H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-27-24-28(30)26-29(31)25-27/h24-26,30-31H,2-23H2,1H3

InChI key

OHTBGMREZYLZQD-UHFFFAOYSA-N

General description

5-Tricosylresorcinol, belonging to the class of alkylresorcinols (ARs), is an important constituent of cereal grains. Alkylresorcinols exhibit anti-inflammatory, anti-tumor and anti-mutagenic properties, as studied from cell culture.

Application

5-Tricosylresorcinol may be used as an analytical reference standard in the following:
  • Quantification of the analyte in seeds using ultra-high performance liquid chromatography electrospray ionization coupled to quadrupole-time-of-flight mass spectrometry (UHPLC-ESI-QTOF-MS).       
  • Quantification of the analyte in Keishibukuryogan-ka-yokuinin Japanese medicine, using blood pharmacokinetic techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UHPLC-ESI-QTOF-MS screening of lignans and other phenolics in dry seeds for human consumption
Ghisoni S, et al.
Journal of functional foods, 34, 229-236 (2017)
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 2 (2012)
Alkylresorcinol composition allows the differentiation of Triticum spp. having different degrees of ploidy
Ziegler UJ, et al.
Journal of Cereal Science, 65, 244-251 (2015)
Oral administration of the Japanese traditional medicine keishibukuryogan-ka-yokuinin decreases reactive oxygen metabolites in rat plasma: Identification of chemical constituents contributing to antioxidant activity
Matsubara Y, et al.
Molecules (Basel), 22(2), 256-256 (2017)
Yingdong Zhu et al.
Bioorganic & medicinal chemistry, 19(13), 3973-3982 (2011-06-11)
We and others have found that wheat bran oil is the active constituent in wheat bran for colon cancer prevention. However, the active components in wheat bran oil are still unknown. Using human colon cancer cells (HCT-116 and HT-29) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service