Skip to Content
Merck
All Photos(2)

Documents

152633

Sigma-Aldrich

Olivetol

95%

Synonym(s):

1,3-Dihydroxy-5-pentylbenzene, 3,5-Dihydroxyamylbenzene, 5-Pentylresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

46-48 °C (lit.)

SMILES string

CCCCCc1cc(O)cc(O)c1

InChI

1S/C11H16O2/c1-2-3-4-5-9-6-10(12)8-11(13)7-9/h6-8,12-13H,2-5H2,1H3

InChI key

IRMPFYJSHJGOPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Olivetol was used as a template molecule in the synthesis of molecularly imprinted polymer (MIP). It was also used as an inhibitor of (S)-mephenytoin 4′-hydroxylase activity of recombinant CYP2C19.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Cascio et al.
British journal of pharmacology, 149(4), 431-440 (2006-09-06)
We have previously reported the development of CB-25 and CB-52, two ligands of CB1 and CB2 cannabinoid receptors. We assessed here their functional activity. The effect of the two compounds on forskolin-induced cAMP formation in intact cells or GTP-gamma-S binding
M Fellermeier et al.
FEBS letters, 427(2), 283-285 (1998-06-02)
A new enzyme, geranylpyrophosphate:olivetolate geranyltransferase (GOT), the first enzyme in the biosynthesis of cannabinoids could be detected in extracts of young leaves of Cannabis sativa. The enzyme accepts geranylpyrophosphate (GPP) and to a lesser degree also nerylpyrophosphate (NPP) as a
E A Formukong et al.
Inflammation, 12(4), 361-371 (1988-08-01)
Two extracts of Cannabis sativa herb, one being cannabinoid-free (ethanol) and the other containing the cannabinoids (petroleum), were shown to inhibit PBQ-induced writhing in mouse when given orally and also to antagonize tetradecanoylphorbol acetate (TPA)-induced erythema of mouse skin when
Rongrong Jiang et al.
Drug metabolism and pharmacokinetics, 28(4), 332-338 (2013-01-16)
The present study investigated the inhibitory effect of cannabidiol (CBD), a major constituent of marijuana, on the catalytic activity of cytochrome P450 2C19 (CYP2C19). (S)-Mephenytoin 4'-hydroxylase activities of human liver microsomes (HLMs) and recombinant CYP2C19 were inhibited by CBD in
R H McClanahan et al.
Journal of natural products, 47(5), 828-834 (1984-09-01)
A study of the biotransformation of olivetol by Syncephalastrum racemosum ATCC 18192 has led to the isolation of three metabolites, which were identified as 4'-hydroxy-olivetol, 3-(3,5-dihydroxyphenyl)-1-propanol, and 3-(3,5-dihydroxyphenyl)-1-propanoic acid. The structures of the isolated metabolites were deduced by comparison of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service