Skip to Content
Merck
All Photos(1)

Key Documents

15-0330

Sigma-Aldrich

3-Indoleacetic acid

SAJ special grade, ≥98.5%

Synonym(s):

Heteroauxin, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
143358
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
grade:
SAJ special grade

grade

SAJ special grade

Assay

≥98.5%

availability

available only in Japan

dilution

(for analytical testing)

mp

165-169 °C (lit.)

SMILES string

OC(=O)Cc1c[nH]c2ccccc12

InChI

1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)

InChI key

SEOVTRFCIGRIMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L O Villacorte et al.
Water research, 73, 216-230 (2015-02-16)
Algal blooms can seriously affect the operation of water treatment processes including low pressure (micro- and ultra-filtration) and high pressure (nanofiltration and reverse osmosis) membranes mainly due to accumulation of algal-derived organic matter (AOM). In this study, the different components
Amir H Ahkami et al.
Planta, 238(3), 499-517 (2013-06-15)
To determine the contribution of polar auxin transport (PAT) to auxin accumulation and to adventitious root (AR) formation in the stem base of Petunia hybrida shoot tip cuttings, the level of indole-3-acetic acid (IAA) was monitored in non-treated cuttings and
Thomas U Greiner et al.
PloS one, 9(11), e110359-e110359 (2014-11-13)
Islet autoimmunity in children who later progress to type 1 diabetes is preceded by dysregulated serum metabolite profiles, but the origin of these metabolic changes is unknown. The gut microbiota affects host metabolism and changes in its composition contribute to
Xinhua Dai et al.
The Journal of biological chemistry, 288(3), 1448-1457 (2012-11-29)
Auxin regulates every aspect of plant growth and development. Previous genetic studies demonstrated that YUCCA (YUC) flavin-containing monooxygenases (FMOs) catalyze a rate-limiting step in auxin biosynthesis and that YUCs are essential for many developmental processes. We proposed that YUCs convert
Kim L Johnson et al.
PloS one, 10(7), e0131103-e0131103 (2015-07-07)
Mitogen-activated dual-specificity MAPK phosphatases are important negative regulators in the MAPK signalling pathways responsible for many essential processes in plants. In a screen for mutants with reduced organ size we have identified a mutation in the active site of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service