Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Indoleacetic acid
Synonym(s):
Heteroauxin, IAA
Empirical Formula (Hill Notation):
C
10
H
9
NO
2
CAS Number:
87-51-4
Molecular Weight:
175.18
EC Number:
201-748-2
Products
Product Category
building blocks (1)
pesticides (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
I3750
3-Indoleacetic acid,
98%
Expand
View Pricing
I2886
3-Indoleacetic acid,
suitable for plant cell culture, crystalline
Expand
View Pricing
45533
3-Indoleacetic acid,
PESTANAL
®
, analytical standard
Expand
View Pricing
15-0330
3-Indoleacetic acid,
SAJ special grade, ≥98.5%
Expand
View Pricing