05-2751
Calcium hypochlorite
granular, SAJ first grade, available chlorine ≥60 %
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Ca(OCl)2
CAS Number:
Molecular Weight:
142.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Recommended Products
grade
SAJ first grade
form
granular
composition
available chlorine, ≥60%
availability
available only in Japan
mp
100 °C (lit.)
density
2.35 g/mL at 25 °C (lit.)
SMILES string
ClO[Ca]OCl
InChI
1S/Ca.2ClO/c;2*1-2/q+2;2*-1
InChI key
ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gursharan Singh et al.
The Journal of general and applied microbiology, 55(4), 283-289 (2009-08-25)
An alkalophilic and halotolerant laccase from gamma-proteobacterium JB catalyzed in high concentrations of organic solvents and various salts. The enzyme retained 80-100% activity in 10% concentration of dimethylsulfoxide (DMSO), ethanol, acetone or methanol; 100, 85 and 50% activity in 20
Peter K Jensen et al.
Journal of health, population, and nutrition, 21(1), 26-31 (2003-05-20)
To evaluate the importance of public-domain transmission of pathogens in drinking-water, an intervention study was carried out by chlorinating the public water-supply system in a village in Pakistan. The water quality improved and reached a geometric mean of 3 Escherichia
F Boukhoubza et al.
Journal of environmental management, 91(1), 124-132 (2009-09-02)
The application of hypochlorite for the removal of soluble COD, phenolic and polyphenolic like compounds, and other organic compounds responsible for the olive mill wastewater (OOWW) colour has been experimentally studied. After the OOWW filtration on a sand column, the
Hudaa Neetoo et al.
International journal of food microbiology, 131(2-3), 218-223 (2009-04-03)
The application of high hydrostatic pressure technology as a seed decontamination technology was evaluated. Alfalfa seeds inoculated with approximately 10(5) CFU/g of Escherichia coli O157:H7 were subjected to oscillatory pressure treatments at 600 MPa and 20 degrees C for up
M L Bari et al.
Journal of food protection, 73(4), 752-757 (2010-04-10)
The majority of the seed sprout-related outbreaks have been associated with Escherichia coli O157:H7 and Salmonella. Therefore, an effective method for inactivating these organisms on the seeds before sprouting is needed. The current pasteurization method for mung beans in Japan
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service