Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium hypochlorite
Synonym(s):
Calcium hypochlorite
Linear Formula:
Ca(OCl)
2
CAS Number:
7778-54-3
Molecular Weight:
142.98
EC Number:
231-908-7
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Reagent Type
Special Grade
Technical (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
211389
Calcium hypochlorite,
technical grade
Expand
View Pricing
8.41799
Calcium hypochlorite,
for synthesis
Expand
View Pricing
05-2751
Calcium hypochlorite,
granular, SAJ first grade, available chlorine ≥60 %
Expand
View Pricing