Skip to Content
Merck
All Photos(1)

Documents

05-1420

Sigma-Aldrich

Octanoic acid

≥96.0%

Synonym(s):

Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747180
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 78 °C)

Assay

≥96.0%

availability

available only in Japan

refractive index

n20/D 1.428 (lit.)

bp

237 °C (lit.)

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

functional group

carboxylic acid

storage temp.

room temp

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

amino acid analysis

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romain Harmancey et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(10), 3966-3978 (2013-07-05)
Diabetic patients with acute myocardial infarction are more likely to die than nondiabetic patients. In the present study we examined the effect of insulin resistance on myocardial ischemia tolerance. Hearts of rats, rendered insulin resistant by high-sucrose feeding, were subjected
Xuan Jiang et al.
Biomacromolecules, 13(9), 2926-2932 (2012-08-09)
When grown in a nonanoic acid-limited chemostat at a dilution rate of 0.25 h(-1), Pseudomonas putida KT2440 produced poly(3-hydroxynonanoate-co-3-hydroxyheptanoate) containing 68 mol % 3-hydroxynonanoate (C9) and 32 mol % 3-hydroxyheptanoate (C7). Under the same conditions, but in the presence of
Hong-Fei Tong et al.
Journal of chromatography. A, 1285, 88-96 (2013-03-12)
Hydrophobic charge-induction chromatography (HCIC) is a novel downstream bioprocessing technology for antibody purification and it has several advantages over traditional purification processes. However, its separation selectivity still needs to be improved. In this work, sodium caprylate (NaCA) was used as
K M Morrill et al.
Journal of dairy science, 95(7), 3987-3996 (2012-06-23)
Our objectives were to evaluate the use of refractometry as a means of estimating immunoglobulin G (IgG) concentration of bovine maternal colostrum (MC) and determine if fractionation of MC using caprylic acid (CA) improved estimates of IgG. Samples (n=85) of
Sara Janssen et al.
PloS one, 7(6), e40168-e40168 (2012-07-07)
Ghrelin is an important regulator of energy--and glucose homeostasis. The octanoylation at Ser(3) is essential for ghrelin's biological effects but the mechanisms involved in the octanoylation are unknown. We investigated whether the gustatory G-protein, α-gustducin, and the free fatty acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service