Skip to Content
Merck
All Photos(1)

Documents

W279900

Sigma-Aldrich

Octanoic acid

≥98%, FG

Synonym(s):

Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2799
Beilstein:
1747180
EC Number:
Council of Europe no.:
10
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.010
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.210
FDA 21 CFR 172.860
FDA 21 CFR 184.1025

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 78 °C)

Assay

≥98%

refractive index

n20/D 1.428 (lit.)

bp

237 °C (lit.)

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; oily; vegetable; waxy; cheesy; rancid

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octanoic acid is a fatty acid mainly found in butter, edible oils and milk of various species.

Application


  • Chemical Characterization of Terpene-Based Hydrophobic Eutectic Solvents and Their Application for Pb(II) Complexation during Solvent Extraction Procedure.: Research investigates the use of octanoic acid in the synthesis of novel terpene-based eutectic solvents, focusing on their application in heavy metal extraction. This study is crucial for environmental chemistry, offering a sustainable alternative for pollutant removal (Suljkanović et al., 2024).

  • Green and Efficient Extraction of Phenolic Components from Plants with Supramolecular Solvents: Experimental and Theoretical Studies.: This research utilizes octanoic acid in the development of supramolecular solvents for the extraction of phenolic compounds from plants. The study focuses on optimizing extraction efficiency and sustainability, enhancing the production of natural antioxidants (Xia et al., 2024).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dielectric properties of edible oils and fatty acids as a function of frequency, temperature, moisture and composition
Lizhi H, et al.
Journal of Food Engineering, 88(2), 151-158 (2008)
Characterization of edible oils, butters and margarines by Fourier transform infrared spectroscopy with attenuated total reflectance.
Safar M, et al.
Journal of the American Oil Chemists' Society, 71(4), 371-377 (1994)
Antibacterial effect of caprylic acid and monocaprylin on major bacterial mastitis pathogens.
Nair MKM, et al.
Journal of Dairy Science, 88(10), 3488-3495 (2005)
Xuan Jiang et al.
Biomacromolecules, 13(9), 2926-2932 (2012-08-09)
When grown in a nonanoic acid-limited chemostat at a dilution rate of 0.25 h(-1), Pseudomonas putida KT2440 produced poly(3-hydroxynonanoate-co-3-hydroxyheptanoate) containing 68 mol % 3-hydroxynonanoate (C9) and 32 mol % 3-hydroxyheptanoate (C7). Under the same conditions, but in the presence of
K M Morrill et al.
Journal of dairy science, 95(7), 3987-3996 (2012-06-23)
Our objectives were to evaluate the use of refractometry as a means of estimating immunoglobulin G (IgG) concentration of bovine maternal colostrum (MC) and determine if fractionation of MC using caprylic acid (CA) improved estimates of IgG. Samples (n=85) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service