Skip to Content
Merck
All Photos(2)

Key Documents

8.02934

Sigma-Aldrich

Diethylene glycol dimethyl ether

(stabilised) for synthesis

Synonym(s):

2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736101
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-924-4
NACRES:
NA.22

vapor density

4.6 (vs air)

Quality Level

vapor pressure

3 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

370 °F

potency

4760 mg/kg LD50, oral (Rat)

expl. lim.

17.4 %

refractive index

n20/D 1.408 (lit.)

pH

7 (20 °C)

bp

162 °C (lit.)

mp

−64 °C (lit.)

transition temp

flash point 51 °C

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.943 - 0.945
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service