Skip to Content
Merck
All Photos(1)

Key Documents

8.02932

Sigma-Aldrich

Diethylene glycol diethyl ether

for synthesis

Synonym(s):

Diethylene glycol diethyl ether, Diethyl diglycol, Bis(2-ethoxyethyl) ether, 2-Ethoxyethyl ether, Diethyl Carbitol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5OCH2CH2)2O
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-963-7
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
189 °C/1013 hPa
vapor pressure:
0.5 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.5 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

174 °C

potency

4970 mg/kg LD50, oral (Rat)
6097 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

bp

189 °C/1013 hPa

mp

-44 °C

transition temp

flash point 71 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCC)CCOCC

InChI

1S/C8H18O3/c1-3-9-5-7-11-8-6-10-4-2/h3-8H2,1-2H3

InChI key

RRQYJINTUHWNHW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.906 - 0.908
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Legal Information

CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service