The weight of this item is approximately 6.5 KG.
Z126195
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Synonym(s):
Thin Layer Chromatography Tank
About This Item
Recommended Products
technique(s)
thin layer chromatography (TLC): suitable
L × H × W
27.0 cm × 26.5 cm × 7.0 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Footnote
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the mass of the item in Kg?
1 answer-
Helpful?
-
-
Bonjour, quelle est la résistance thermique et la nature du verre de cette cuve? Je souhaiterait d'utiliser à chaud et chauffée par plaque chauffante. Merci
1 answer-
The temperature resistance of this glass has not been determined. This tank is seamless and made of borosilicate glass, which typically has a high temperature resistance. Most sources state that this glass is resistant to a range of 300-500°C. However, this product has not been tested under high heat conditions, which are often incompatible with TLC reagents.
Helpful?
-
-
Hi, is the "Rectangular TLC developing tanks, complete" made with the glass?
1 answer-
Yes, this product is made of borosilicate glass.
Helpful?
-
-
How thick is the glass?
1 answer-
The thickness of the glass may vary from 3/8 to 3/4 inches.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service