Skip to Content
Merck
All Photos(1)

Key Documents

CDS006238

Sigma-Aldrich

PhenylAc-Gln-OH

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O4
Molecular Weight:
264.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

NC(=O)CC[C@H](NC(=O)Cc1ccccc1)C(O)=O

InChI

1S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1

InChI key

JFLIEFSWGNOPJJ-JTQLQIEISA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadezda V Khodorova et al.
Molecular nutrition & food research, 63(1), e1700834-e1700834 (2018-02-23)
The impact of meat consumption on human health is widely examined in nutritional epidemiological studies, especially due to the connection between the consumption of red and processed meat and the risk of colon cancer. Food questionnaires do not assess the
Tiffany A Freed et al.
Clinical chemistry, 65(3), 406-418 (2019-01-17)
Clinical practice guidelines recommend estimation of glomerular filtration rate (eGFR) using validated equations based on serum creatinine (eGFRcr), cystatin C (eGFRcys), or both (eGFRcr-cys). However, when compared with the measured GFR (mGFR), only eGFRcr-cys meets recommended performance standards. Our goal
Jing Xu et al.
Talanta, 192, 160-168 (2018-10-24)
The pseudo-targeted metabolomics approach was developed recently which combined the advantages of untargeted and targeted analysis. However, the current pseudo-targeted analysis method has limitations due to the technical characteristics. In this study, a novel metabolic pathway-based pseudo-targeted approach was proposed
Sergio Lario et al.
Scientific reports, 7(1), 17774-17774 (2017-12-21)
Gastric carcinogenesis is a multifactorial process described as a stepwise progression from non-active gastritis (NAG), chronic active gastritis (CAG), precursor lesions of gastric cancer (PLGC) and gastric adenocarcinoma. Gastric cancer (GC) 5-year survival rate is highly dependent upon stage of
G van Straten et al.
Scientific reports, 7(1), 13112-13112 (2017-10-17)
Urea cycle enzyme deficiency (UCED) patients with hyperammonemia are treated with sodium benzoate (SB) and sodium phenylacetate (SPA) to induce alternative pathways of nitrogen excretion. The suggested guidelines supporting their use in the management of hyperammonemia are primarily based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service