Skip to Content
Merck
All Photos(1)

Key Documents

930954

Sigma-Aldrich

Aluminum nitrate nonahydrate

99.99% (trace metals basis)

Synonym(s):

Aluminum nitrate hydrate, Nitric acid aluminum salt nonahydrate, Aluminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
375.13
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
form:
powder
solubility:
acetone: soluble
alcohols: soluble
water: soluble (highly soluble)

description

(2.5 - 3.5 at 50 g/L at 25 °C)

Quality Level

Assay

99.99% (trace metals basis)

form

powder

impurities

≤150 ppm (trace metals analysis)

mp

73 °C (lit.)

solubility

acetone: soluble
alcohols: soluble
water: soluble (highly soluble)

density

1.72 g/cm3

SMILES string

O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum nitrate nonahydrate is a white crystalline solid and highly soluble in water. It is a strong oxidizing agent and widely used in field of catalysis, solar cells, semiconductors, and metal coatings.

Application

Aluminum nitrate nonahydrate can be used :
  • As a precursor to prepare Al-ion electrolyte for dye-sensitized solar cells (DSSCs). It enhances photoconversion efficiency of solar cells.
  • As a dopant to prepare Al-ZnO thin films for solar cells by sol-gel method.
  • As a starting material to fabricate NiAl layered hydroxide electrode with high capacity and more stable cycling ability, for Li-ion batteries.
  • To prepare α-alumina fibers by sol-gel processing. These dense fibers are used in the fabrication of catalytic supports, electronic devices, anticorrosion coating, and medical implants.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Operando Identification of Geometrical-Site-Dependent Water Oxidation Activity of Spinel Co3O4.
Wang, H., Hung, S., et al.
Journal of the American Chemical Society, 138, 36-39 (2016)
Magnetic polydopamine decorated with Mg-Al LDH nanoflakes as a novel bio-based adsorbent for simultaneous removal of potentially toxic metals and anionic dyes.
Li, J., Fan, Q., et al.
Journal of Material Chemistry A, 4, 1737-1746 (2016)
Biotemplated Hierarchical Nanostructure of Layered Double Hydroxides with Improved Photocatalysis Performance.
Zhao, Y., Wei, M., et al.
ACS Nano, 3, 4009-4016 (2009)
Improving thermal and electrochemical performances of LiCoO2 cathode at high cut-off charge potentials by MF3 (M=Ce, Al) coating.
Aboulaich, A., Ouzaouit, K., et al.
Materials Research Bulletin, 73, 362-368 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service