Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum nitrate nonahydrate
Synonym(s):
Aluminium nitrate
Linear Formula:
Al(NO
3
)
3
· 9H
2
O
CAS Number:
7784-27-2
Molecular Weight:
375.13
EC Number:
236-751-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
237973
Aluminum nitrate nonahydrate,
ACS reagent, ≥98%
Expand
View Pricing
229415
Aluminum nitrate nonahydrate,
99.997% trace metals basis
Expand
View Pricing
930954
Aluminum nitrate nonahydrate,
99.99% (trace metals basis)
Expand
View Pricing
01-2790
Aluminum nitrate nonahydrate,
JIS special grade, ≥98.0%
Expand
View Pricing