Skip to Content
Merck
All Photos(1)

Documents

371475

Sigma-Aldrich

2,2-Dimethylcyclopentanone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C5H6(=O)
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.433 (lit.)

bp

143-145 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CCCC1=O

InChI

1S/C7H12O/c1-7(2)5-3-4-6(7)8/h3-5H2,1-2H3

InChI key

FTGZMZBYOHMEPS-UHFFFAOYSA-N

General description

2,2-Dimethylcyclopentanone is a ketone. Synthesis of various C-2 substituted vitamin D derivatives with a 2,2-dimethylcyclopentanone unit in side chains has been reported. Regioselective synthesis of 2,2-dimethylcyclopentanone, via its enolate precursor regioselectively obtained using the 2-pyrrolidone magnesium salt, has been reported.

Application

2,2-Dimethylcyclopentanone enolate may be used as starting reagent in the enantioselective synthesis of chiral phosphines belonging to the P-aryl-2-phosphabicyclo[3.3.0]octane family (PBO). It may be used in the synthesis of:
  • 2,6,6-trimethyl-2-azaspiro[4.4]nonane-1,3-dione, a spirosuccinimide moiety of asperparaline A
  • novel spiropentanopyrrolizidine oxime alkaloids, namely 2′,3′,5′,6′,7′,7a′-hexahydro-2,2-dimethylspirocyclopentane-1
  • δ,δ-dimethyl-δ-valerolactone, via Baeyer-Villiger oxidation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of catalpalactone.
Lane KJ and Pinder AR.
The Journal of Organic Chemistry, 47(16), 3171-3172 (1982)
Synthesis of pyrrolizidine oximes 222 and 236: Novel alkaloids of a dendrobatid poison frog.
Rutchinson KD, et al.
Tetrahedron, 50(21), 6129-6136 (1994)
Edwin Vedejs et al.
Journal of the American Chemical Society, 125(14), 4166-4173 (2003-04-03)
A new class of chiral phosphines belonging to the P-aryl-2-phosphabicyclo[3.3.0]octane family (PBO) has been prepared by enantioselective synthesis starting from lactate esters and 2,2-dimethylcyclopentanone enolate 5. A selective enolate alkylation method has been developed for preparation of 9 and 10
Regioselective Synthesis of 2, 2-Dimethylcyclopentanone Using 2-Pyrrolidone Magnesium Salt as Electrogenerated Base.
Bonafoux D, et al.
Synthetic Communications, 28(1), 93-98 (1998)
S Tanimori et al.
Bioscience, biotechnology, and biochemistry, 64(8), 1758-1760 (2000-09-19)
2,6,6-Trimethyl-2-azaspiro[4.4]nonane-1,3-dione (9), a spirosuccinimide moiety of asperparaline A (1), was synthesized by starting from 2,2-dimethylcyclopentanone (4) via trinitrile 6 in five steps in a moderate yield. This conversion establishes a model study for synthesis of the spirosuccinimide moiety of asperparaline

Articles

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service