Skip to Content
Merck
All Photos(1)

Key Documents

8.02670

Sigma-Aldrich

Cyclopentanone

for synthesis

Synonym(s):

Cyclopentanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O
CAS Number:
Molecular Weight:
84.12
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-435-9
NACRES:
NA.22
grade:
for synthesis
bp:
131 °C/1013 hPa
vapor pressure:
11.5 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

11.5 hPa ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

445 °C

potency

>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-10.8 % (v/v)

dilution

(for synthesis)

bp

131 °C/1013 hPa

mp

-58 °C

transition temp

flash point 27 °C

solubility

slightly soluble 9.18 g/L

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C1CCCC1

InChI

1S/C5H8O/c6-5-3-1-2-4-5/h1-4H2

InChI key

BGTOWKSIORTVQH-UHFFFAOYSA-N

Application


  • Cyclopentanone as a catalyst in stereoselective synthesis: Research highlighted the use of cyclopentanone in enhancing substrate stereoselectivity during the isothiourea-catalyzed conjugate addition of cyclic alpha-substituted beta-ketoesters to alpha,beta-unsaturated aryl esters, providing a pathway for precision in chemical synthesis. (Yuan et al., 2023).

  • Role in the synthesis of metal complexes: Cyclopentanone contributed to the synthesis, structural determination, and reactivity study of complex metal systems such as trindane and truxene, underscoring its utility in advanced organic synthesis and materials science. (Lock et al., 2023).

  • Impact on the properties of fluorinated derivatives: A theoretical study involving cyclopentanone examined the molecular properties of fluorinated derivatives, highlighting its importance in the development of high-performance solvents and pharmaceutical intermediates. (Tshepelevitsh et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.947 - 0.949
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service