Skip to Content
Merck
All Photos(1)

Documents

244414

Sigma-Aldrich

1-Heptyne

98%

Synonym(s):

1-n-Heptyne, Amylacetylene, Pentylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C≡CH
CAS Number:
Molecular Weight:
96.17
Beilstein:
1733518
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010410
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

93.1 mmHg ( 37.7 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.408 (lit.)

bp

99-100 °C (lit.)

mp

−81 °C (lit.)

density

0.733 g/mL at 25 °C (lit.)

SMILES string

CCCCCC#C

InChI

1S/C7H12/c1-3-5-7-6-4-2/h1H,4-7H2,2H3

InChI key

YVXHZKKCZYLQOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The hydrogenation of 1-heptyne with palladium, platinum and ruthenium supported on activated carbon as catalysts, was studied.

Application

1-Heptyne was used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F

Flash Point(C)

-10 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Loreau et al.
Chemistry and physics of lipids, 124(2), 135-145 (2003-06-24)
To study the metabolic fate of conjugated linoleic acid isomers, we synthesized, in seven steps, from 1-heptyne, (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid, (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid, and their [1-(14)C]-analogs. In the case of (6Z,10E,12Z)-octadecatrienoic acid, a series of palladium-catalyzed cross-coupling reactions between 1-heptyne and (E)-1,2-dichloro-ethene
Cecilia R Lederhos et al.
TheScientificWorldJournal, 2013, 528453-528453 (2013-12-19)
Palladium, platinum, and ruthenium supported on activated carbon were used as catalysts for the selective hydrogenation of 1-heptyne, a terminal alkyne. All catalysts were characterized by temperature programmed reduction, X-ray diffraction, transmission electron microscopy, and X-ray photoelectron spectroscopy. TPR and
Chloë L Wright et al.
Applied and environmental microbiology, 86(9) (2020-02-23)
Ammonia monooxygenase (AMO) is a key nitrogen-transforming enzyme belonging to the same copper-dependent membrane monooxygenase family (CuMMO) as the particulate methane monooxygenase (pMMO). The AMO from ammonia-oxidizing archaea (AOA) is very divergent from both the AMO of ammonia-oxidizing bacteria (AOB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service