All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
vapor pressure
1.1 psi ( 37.7 °C)
Assay
99%
refractive index
n20/D 1.392 (lit.)
bp
108 °C (lit.)
density
0.694 g/mL at 25 °C (lit.)
SMILES string
CC(C)CCC(C)C
InChI
1S/C8H18/c1-7(2)5-6-8(3)4/h7-8H,5-6H2,1-4H3
InChI key
UWNADWZGEHDQAB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2,5-Dimethylhexane has been used to study excess entropy scaling relationships for analyzing its kinetic properties using molecular simulation.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
78.8 °F - closed cup
Flash Point(C)
26 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ravi Chopra et al.
The journal of physical chemistry. B, 114(49), 16487-16493 (2010-11-26)
We use molecular simulation to study the ability of excess entropy scaling relationships to describe the kinetic properties of four hydrocarbon isomers: n-octane, 2,2-dimethylhexane, 2,5-dimethylhexane, and 3-methyl-3-ethylpentane. Four dynamic properties are considered: translational and rotational diffusivities, a characteristic relaxation time
Barbara Nozière et al.
Angewandte Chemie (International ed. in English), 58(39), 13976-13982 (2019-07-31)
The autoxidation of organic peroxy radicals (RO2 ) into hydroperoxy-alkyl radicals (QOOH), then hydroperoxy-peroxy radicals (HOOQO2 ) is now considered to be important in the Earth's atmosphere. To avoid mechanistic uncertainties these reactions are best studied by monitoring the radicals.
Michael A Wheeler et al.
Cell, 176(3), 581-596 (2019-01-22)
Genome-wide studies have identified genetic variants linked to neurologic diseases. Environmental factors also play important roles, but no methods are available for their comprehensive investigation. We developed an approach that combines genomic data, screens in a novel zebrafish model, computational
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service