Skip to Content
Merck
All Photos(3)

Key Documents

213365

Sigma-Aldrich

Selenium dioxide

ReagentPlus®, powder, 99.8% trace metals basis

Synonym(s):

NSC 56753, Selenium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeO2
CAS Number:
Molecular Weight:
110.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 157 °C)

Quality Level

product line

ReagentPlus®

Assay

99.8% trace metals basis

form

powder

color

white

mp

315 °C (subl.) (lit.)

SMILES string

O=[Se]=O

InChI

1S/O2Se/c1-3-2

InChI key

JPJALAQPGMAKDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mild oxidizing reagent for allylic olefins and acetylenes. Conversion of 1,3-diketones to 1,2,3-triones

Other Notes

Material may contain dark crystals which are unavoidable and do not impact product quality

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asian Journal of Chemistry, 19, 4107-4107 (2007)
Tetrahedron Letters, 34, 2979-2979 (1993)
Erik A A Wallén et al.
Organic letters, 9(3), 389-391 (2007-01-26)
[reaction: see text] An efficient synthetic route to Cbz-protected 3-aminomethyl-2-aryl-8-bromo-6-chlorochromones has been developed. 3-Aryl-1-(3-bromo-5-chloro-2-hydroxyphenyl)-2-propen-1-one or 2-aryl-8-bromo-6-chlorochroman-4-one could be reacted under Mannich conditions yielding 2-aryl-8-bromo-6-chloro-3-methylenechroman-4-one, which was further converted to the target compound via an aza-Michael reaction followed by an SeO(2)
Haiying Luo et al.
Colloids and surfaces. B, Biointerfaces, 94, 304-308 (2012-03-02)
In vitro antiproliferative effects of selenium nanoparticles (nanoSe(0), 10-40 μmol/L) on HeLa (human cervical carcinoma) cells and MDA-MB-231 (human breast carcinoma) cells were examined by optical microscopic inspection and MTT assay in the present study. The nanoSe(0) effectively inhibited the
Yu-Wei Chen et al.
Chemosphere, 74(8), 1079-1084 (2008-12-09)
The possible abiotic reduction of selenite to form elemental Se was studied under controlled conditions in the presence of ferrous iron. The reduction of selenite and formation of Se (0) was found to be a surface-mediated reaction by iron oxides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service