Skip to Content
Merck
All Photos(1)

Key Documents

459496

Sigma-Aldrich

9-Borabicyclo[3.3.1]nonane solution

0.4 M in hexanes

Synonym(s):

9-BBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15B
CAS Number:
Molecular Weight:
122.02
Beilstein:
605509
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reagent type: reductant

concentration

0.4 M in hexanes

bp

68-70 °C

density

0.691 g/mL at 25 °C

SMILES string

B1C2CCCC1CCC2

InChI

1S/C8H15B/c1-3-7-5-2-6-8(4-1)9-7/h7-9H,1-6H2/t7-,8+

InChI key

FEJUGLKDZJDVFY-OCAPTIKFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Protecting group for alkenes†

Reactant for:
  • Linear SPPS synthesis of ubiquitin derivatives
  • Copper-catalyzed cross-coupling reactions of organoboron compounds with primary alkyl halides and pseudohalides
  • Intramolecular insertion of alkenes into palladium-nitrogen bonds
  • Preparation of (phosphonoacetyl)ornithine to study effect on arginine biosynthetic genes in yeast
  • Hetero-Diels-Alder reaction for synthesis of spirocyclic alkaloids

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jefferson D Revell et al.
Organic letters, 7(5), 831-833 (2005-02-25)
1,5-Cyclooctadiene was deprotonated under LICKOR conditions and reacted with Merrifield resin to afford an immobilized cyclooctadiene in high yield. This polymer is effective as a halogen scavenger, while hydroboration leads to a supported 9-BBN analogue. The latter exhibits similar regioselectivity
Yi Luan et al.
Organic letters, 13(9), 2510-2513 (2011-04-09)
Diazo esters, arylboranes, and carbamoyl imines undergo a 3-component Mannich condensation reaction. Catalyzed by Cu(II) salts, the reaction affords the corresponding isocyanate Mannich product: one that can be easily trapped in situ by other nucleophiles. The Mannich condensation corresponds to
A Ganesan
Mini reviews in medicinal chemistry, 6(1), 3-10 (2006-02-07)
Solid-phase synthesis is a powerful tool for achieving high-throughput chemistry. This review discusses recent diverse examples from my group: the solid-phase synthesis of unsymmetrical guanidines, polymer-supported versions of cyclooctadiene and 9-BBN, a triflate-like linker, the synthesis of tetrahydro-beta-carbolines by the
Rajesh Sardar et al.
Journal of the American Chemical Society, 133(21), 8179-8190 (2011-05-10)
We report a spectroscopic and microscopic investigation of the synthesis of gold nanoparticles (AuNPs) with average sizes of less than 5 nm. The slow reduction and AuNP formation processes that occur by using 9-borabicyclo[3.3.1]nonane (9-BBN) as a reducing agent enabled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service