Skip to Content
Merck
All Photos(2)

Documents

202967

Sigma-Aldrich

Calcium nitrate hydrate

99.997% trace metals basis

Synonym(s):

Nitric acid calcium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2·xH2O
CAS Number:
Molecular Weight:
164.09 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.997% trace metals basis

form

crystals and lumps

impurities

≤35.0 ppm Trace Metal Analysis

mp

45 °C (lit.)

SMILES string

O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

JJIQGEZLLWXYKV-UHFFFAOYSA-N

General description

Calcium nitrate hydrate can be used as a precursor to synthesize Ca-based nanopowder via a sol-gel process. It can also be used to prepare aerosol for cloud condensation activity.

Application

  • Crack-Repair Efficiency: Evaluating the efficiency of calcium nitrate in the self-healing of cracks in steel fiber-reinforced concrete beams (Milla et al., 2019).

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amin Sadeghizadeh et al.
Journal of environmental management, 232, 342-353 (2018-11-30)
In the current study, the prediction efficiency of lead adsorption by highly functional nanocomposite adsorbent of hydroxyapatite (HAp)/chitosan using ANFIS system was investigated. In this regard, the nanocomposite was applied in order to investigate the lead adsorption capacity. The operational
Fatma S Abdel-Salam et al.
International journal of pharmaceutics, 580, 119213-119213 (2020-03-14)
In-situ forming implants receive great attention for repairing serious bone injuries. The aim of the present study was to prepare novel chitosan in-situ forming implants (CIFI) loaded with bioactive glass nanoparticles and/or raloxifene hydrochloride (RLX). Incorporating raloxifene hydrochloride (RLX) as
Bo Wu et al.
Journal of colloid and interface science, 527, 195-201 (2018-05-25)
Poly(carboxylate ether)-based (PCE) superplasticizers consist of a carboxylic acid backbone and grafted poly(ethylene glycol) (PEG) side chains. Ca2+ ion bridging mechanism is commonly purported to control PCE's adsorption on negatively charged cement particle surfaces in cement suspension, thus PCE was
Justine C Lee et al.
The Journal of craniofacial surgery, 26(6), 1992-1996 (2015-07-07)
Osseous defects of the craniofacial skeleton occur frequently in congenital, posttraumatic, and postoncologic deformities. The field of scaffold-based bone engineering emerged to address the limitations of using autologous bone for reconstruction of such circumstances. In this work, the authors evaluate
Xiaoyan Ren et al.
Science advances, 5(6), eaaw4991-eaaw4991 (2019-06-18)
The instructive capabilities of extracellular matrix-inspired materials for osteoprogenitor differentiation have sparked interest in understanding modulation of other cell types within the bone regenerative microenvironment. We previously demonstrated that nanoparticulate mineralized collagen glycosaminoglycan (MC-GAG) scaffolds efficiently induced osteoprogenitor differentiation and

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service