Skip to Content
Merck
All Photos(3)

Documents

131776

Sigma-Aldrich

Sarcosine

98%, suitable for cell culture

Synonym(s):

N-Methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
1699442
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product name

Sarcosine, 98%

Assay

98%

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

color

white

mp

208-212 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CNCC(O)=O

InChI

1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)

InChI key

FSYKKLYZXJSNPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dandan Yao et al.
Communications biology, 4(1), 933-933 (2021-08-21)
During duration spaceflight, or after their return to earth, astronauts have often suffered from gait instability and cerebellar ataxia. Here, we use a mouse model of hindlimb unloading (HU) to explore a mechanism of how reduced hindlimb burden may contribute
D S Lim et al.
Journal of the American College of Cardiology, 38(4), 1175-1180 (2001-10-05)
The goal of this study was to identify genes upregulated in the heart in human patients with hypertrophic cardiomyopathy (HCM). Hypertrophic cardiomyopathy is a genetic disease caused by mutations in contractile sarcomeric proteins. The molecular basis of diverse clinical and
Eri Hara et al.
Biochimica et biophysica acta, 1830(8), 4046-4052 (2013-04-03)
Nanoparticle of Lactosome, which is composed of poly(l-lactic acid)-base depsipeptide with diameter of 35nm, accumulates in solid tumors by the enhanced permeability and retention (EPR) effect. However, a pharmacokinetic alteration of Lactosome was observed when Lactosome was repeatedly administered. This
J R Bales et al.
Clinical chemistry, 30(10), 1631-1636 (1984-10-01)
Acetaminophen and its glucuronide, sulfate, N-acetyl-L-cysteinyl, and L-cysteinyl metabolites can be rapidly detected by 1H NMR spectroscopy of intact, untreated human urine. Study of the time course of excretion of these metabolites in five clinically normal men after ingestion of
Simone J P M Eussen et al.
The British journal of nutrition, 110(2), 363-374 (2012-12-12)
Different lifestyle patterns across Europe may influence plasma concentrations of B-vitamins and one-carbon metabolites and their relation to chronic disease. Comparison of published data on one-carbon metabolites in Western European regions is difficult due to differences in sampling procedures and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service