Merck
  • Urinary excretion of acetaminophen and its metabolites as studied by proton NMR spectroscopy.
Urinary excretion of acetaminophen and its metabolites as studied by proton NMR spectroscopy.
Clinical chemistry (1984-10-01)
J R Bales, P J Sadler, J K Nicholson, J A Timbrell
ABSTRACT
Acetaminophen and its glucuronide, sulfate, N-acetyl-L-cysteinyl, and L-cysteinyl metabolites can be rapidly detected by 1H NMR spectroscopy of intact, untreated human urine. Study of the time course of excretion of these metabolites in five clinically normal men after ingestion of the usual 1-g therapeutic dose of the drug showed that the mean 24-h excretion of the drug and these metabolites as determined by NMR was 77.3% of the dose. Respective relative proportions of the above metabolites were 49.9%, 37.6%, 3.0%, and 9.5% (L-cysteinyl plus free drug). Excretion of some other metabolites in urine, including creatinine, citrate, hippurate, and sarcosine was measured concurrently. Excretion of creatinine and sarcosine was closely correlated.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Sarcosine, BioXtra