MilliporeSigma
  • Activation of AMP-Activated Protein Kinase by Adenine Alleviates TNF-Alpha-Induced Inflammation in Human Umbilical Vein Endothelial Cells.
Activation of AMP-Activated Protein Kinase by Adenine Alleviates TNF-Alpha-Induced Inflammation in Human Umbilical Vein Endothelial Cells.
PloS one (2015-11-07)
Yi-Fang Cheng, Guang-Huar Young, Jiun-Tsai Lin, Hyun-Hwa Jang, Chin-Chen Chen, Jing-Yi Nong, Po-Ku Chen, Cheng-Yi Kuo, Shao-Hsuan Kao, Yao-Jen Liang, Han-Min Chen
ABSTRACT
The AMP-activated protein kinase (AMPK) signaling system plays a key role in cellular stress by repressing the inflammatory responses induced by the nuclear factor-kappa B (NF-κB) system. Previous studies suggest that the anti-inflammatory role of AMPK involves activation by adenine, but the mechanism that allows adenine to produce these effects has not yet been elucidated. In human umbilical vein endothelial cells (HUVECs), adenine was observed to induce the phosphorylation of AMPK in both a time- and dose-dependent manner as well as its downstream target acetyl Co-A carboxylase (ACC). Adenine also attenuated NF-κB targeting of gene expression in a dose-dependent manner and decreased monocyte adhesion to HUVECs following tumor necrosis factor (TNF-α) treatment. The short hairpin RNA (shRNA) against AMPK α1 in HUVECs attenuated the adenine-induced inhibition of NF-κB activation in response to TNF-α, thereby suggesting that the anti-inflammatory role of adenine is mediated by AMPK. Following the knockdown of adenosyl phosphoribosyl transferase (APRT) in HUVECs, adenine supplementation failed to induce the phosphorylation of AMPK and ACC. Similarly, the expression of a shRNA against APRT nullified the anti-inflammatory effects of adenine in HUVECs. These results suggested that the role of adenine as an AMPK activator is related to catabolism by APRT, which increases the cellular AMP levels to activate AMPK.
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Adenine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human APRT
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
SAFC
L-Glutamine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Pricing and availability is not currently available.