Merck
  • Multi-residue determination of 115 veterinary drugs and pharmaceutical residues in milk powder, butter, fish tissue and eggs using liquid chromatography-tandem mass spectrometry.
Multi-residue determination of 115 veterinary drugs and pharmaceutical residues in milk powder, butter, fish tissue and eggs using liquid chromatography-tandem mass spectrometry.
Analytica chimica acta (2015-06-21)
Marilena E Dasenaki, Nikolaos S Thomaidis
ABSTRACT
A simple and sensitive multi-residue method for the determination of 115 veterinary drugs and pharmaceuticals, belonging in more than 20 different classes, in butter, milk powder, egg and fish tissue has been developed. The method involves a simple generic solid-liquid extraction step (solvent extraction, SE) with 0.1% formic acid in aqueous solution of EDTA 0.1% (w/v)-acetonitrile (ACN)-methanol (MeOH) (1:1:1, v/v) with additional ultrasonic-assisted extraction. Precipitation of lipids and proteins was promoted by subjecting the extracts at very low temperature (-23°C) for 12h. Further cleanup with hexane ensures fat removal from the matrix. Analysis was performed by liquid chromatography coupled with electrospray ionization and tandem mass spectrometry (LC-ESI-MS/MS). Two separate runs were performed for positive and negative ionization in multiple reaction monitoring mode (MRM). Particular attention was devoted to extraction optimization: different sample-to-extracting volume ratios, different concentrations of formic acid in the extraction solvent and different ultrasonic extraction temperatures were tested in butter, egg and milk powder samples. The method was also applied in fish tissue samples. It was validated, on the basis of international guidelines, for all four matrices. Quantitative analysis was performed by means of standard addition calibration. For over 80% of the analytes, the recoveries were between 50% and 120% in all matrices studied, with RSD values in the range of 1-18%. Limits of detection (LODs) and quantification (LOQs) ranged from 0.008 μg kg(-1) (oxfendazole in butter) to 3.15 μg kg(-1) (hydrochlorthiazide in egg). The evaluated method provides reliable screening, quantification, and identification of 115 veterinary drug and pharmaceutical residues in foods of animal origin and has been successfully applied in real samples.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles