Skip to Content
Merck
All Photos(2)

Key Documents

516961

Sigma-Aldrich

Ammonium formate

≥99.995% trace metals basis

Synonym(s):

Formic acid ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$118.00

$118.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$118.00

About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
Beilstein:
3625095
EC Number:
MDL number:
UNSPSC Code:
12352107
eCl@ss:
39021903
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
form:
crystalline

$118.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥99.995% trace metals basis

form

crystalline

impurities

≤50.0 ppm Trace Metal Analysis

mp

119-121 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammoniumformate is a derivative of formic acid widely used as a hydrogen transfer reagentand formylating agent. It is also used in the preparation and storage of formicacid.

Application

Ammonium formate can be used as:
  • Hydrogen source in reduction of aromatic nitro compounds to corresponding amino derivatives in presence of Cu nanoparticles as catalyst.
  • Formylating agent for synthesis of N-formyl amino acid esters.
  • An OH radical scavenger in the preparation of nanocrystalline uranium and/or thorium oxides.

Features and Benefits

  • Inexpensive and readily available
  • Stable and non-toxic
  • Can be used in conjugation with Pd/C andRaney-Nickel catalyst.
  • Can be easily separated from the reaction mixture

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 1241-1241 (2007)
Yuchen He et al.
Analytical chemistry, 93(9), 4217-4222 (2021-02-23)
Mass spectrometry (MS) serves as the centerpiece technology for proteome, lipidome, and metabolome analysis. To gain a better understanding of the multifaceted networks of myriad regulatory layers in complex organisms, integration of different multiomic layers is increasingly performed, including joint
Nigora Mukhamedova et al.
Bio-protocol, 10(13), e3670-e3670 (2021-03-05)
Lipid rafts are distinct liquid-ordered domains of plasma membranes of most eukaryotic cells providing platform for signaling pathways. Lipid composition of rafts is critical for their structural integrity and for regulation of signaling pathways originating from rafts. Here we provide
Katrin A Becker et al.
Journal of neurochemistry, 154(6), 662-672 (2020-02-15)
A major dose-limiting side effect of docetaxel chemotherapy is peripheral neuropathy. Patients' symptoms include pain, numbness, tingling and burning sensations, and motor weakness in the extremities. The molecular mechanism is currently not understood, and there are no treatments available. Previously
Aaron G Wexler et al.
Cell reports, 36(10), 109683-109683 (2021-09-09)
Clostridioides difficile is the leading cause of nosocomial intestinal infections in the United States. Ingested C. difficile spores encounter host bile acids and other cues that are necessary for germinating into toxin-producing vegetative cells. While gut microbiota disruption (often by antibiotics)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service