Skip to Content
Merck
All Photos(1)

Documents

679186

Sigma-Aldrich

Acetonitrile solution

suitable for HPLC, contains 5 % (v/v) water, 0.1 % (v/v) formic acid, 0.05 % (w/v) ammonium formate

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

product name

Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC

vapor density

1.41 (vs air)

Quality Level

vapor pressure

72.8 mmHg

form

liquid

contains

0.05 % (w/v) ammonium formate
0.1 % (v/v) formic acid
5 % (v/v) water

expl. lim.

8 % (lit.)

technique(s)

HPLC: suitable

bp

81-82 °C (lit.)

mp

-48 °C (lit.)

density

0.782 g/mL at 25 °C
0.807 g/mL at 25 °C

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

LC/MS suitability tested

Application


  • Detection of Amino Acids via Chemiluminescence: Acetonitrile is employed in the detection of α-amino acids using peroxyoxalate chemiluminescence. A study demonstrated the use of a mixed solution of water, acetonitrile, and ethyl acetate for detecting amino acids, highlighting its efficacy in analytical applications. This method enhances the sensitivity and accuracy of amino acid detection, making it crucial for biochemical and clinical analyses (Kan et al., American Journal of Analytical Chemistry, 2020).

  • Redox Properties of N-Hydroxyimides: Acetonitrile mixed with sulfuric acid is used to study the redox properties of N-hydroxyimides, acting as a hydrogen atom transfer (HAT) mediator for alcohol oxidation. This application is significant in understanding the electrochemical behavior of organic compounds and developing new catalytic processes for oxidation reactions (Kishioka, Journal of Electroanalytical Chemistry, 2023).

  • Electrode Reactions in Catalytic Mediation: Acetonitrile solution is used in investigating the electrode reactions of N-hydroxyphthalimide in sulfuric acid. This study focuses on its role as a catalytic mediator for alcohol oxidation, providing insights into the electrochemical mechanisms involved. The findings contribute to the development of efficient catalytic systems for organic transformations (Kishioka, Journal of Electroanalytical Chemistry, 2022).

  • Excitation of Europium Complexes: Acetonitrile solution is utilized in the interaction of Eu(FOD)3 with xenon difluoride to study ligand-assisted excitation of Eu(III). This research explores the photophysical properties of europium complexes, which are essential for developing luminescent materials for various applications, including lighting and display technologies (Masyagutova et al., Journal of Fluorine Chemistry, 2024).


Preparation Note

Product filtered through a 0.2 μm filter

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

36.0 °F - closed cup

Flash Point(C)

2.22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kangmin Chon et al.
Water research, 81, 388-397 (2015-07-05)
In this study, the changes in UV absorbance at 254 nm (UVA254) and electron donating capacity (EDC) were investigated as surrogate indicators for assessing removal of micropollutants and bromate formation during ozonation of wastewater effluents. To measure the EDC, a novel
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic
Maria de la Paz Celorio-Mancera et al.
Insect biochemistry and molecular biology, 61, 34-45 (2015-05-03)
More than half of the proteome from mandibular glands in caterpillars is represented by chemosensory proteins. Based on sequence similarity, these proteins are putative transporters of ligands to gustatory receptors in sensory organs of insects. We sought to determine whether
Fang Xu et al.
The Plant cell, 27(5), 1547-1562 (2015-05-15)
Nod-like receptors (NLRs) serve as immune receptors in plants and animals. The stability of NLRs is tightly regulated, though its mechanism is not well understood. Here, we show the crucial impact of N-terminal acetylation on the turnover of one plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service