Merck
  • Molecularly imprinted polymers as a tool for the study of the 4-ethylphenol metabolic pathway in red wines.
Molecularly imprinted polymers as a tool for the study of the 4-ethylphenol metabolic pathway in red wines.
Journal of chromatography. A (2015-08-11)
Deiene Garcia, Alberto Gomez-Caballero, Antonio Guerreiro, M Aranzazu Goicolea, Ramon J Barrio
ABSTRACT
A molecularly imprinted polymer (MIP) based methodology is described here for the determination of compounds that belong to the 4-ethylphenol (4EP) metabolic pathway in red wines. To this end, two MIP materials have been developed: a 4EP MIP as a class-selective material to extract phenols that belong to the 4EP metabolic pathway and a coumaric acid (CA) imprinted polymer as a MIP-based stationary phase capable of selectively separating these phenols on HPLC analysis, obtaining clean chromatograms. 4-vinyl pyridine and ethylene glycol dimethacrylate were respectively used as functional monomer and cross-linker for both MIPs. Once polymer compositions were optimised, the 4EP MIP was packed into SPE cartridges for wine sample clean-up and CA MIP was packed into HPLC columns to chromatographically separate the compounds present in the eluates obtained after SPE extraction. The accuracy of the proposed method was tested spiking wine samples with known concentrations of target compounds and subsequently, analytes were quantified by the standard addition method. Registered mean recoveries ranged from 95.2 to 109.2% and RSD values were below 10% in most cases. The described methodology was found to be suitable for the selective extraction and quantification of the compounds that belong to the 4EP metabolic pathway in red wines with minimal matrix effects and could be undoubtedly exploited to monitor 4EP and its precursors in wines.
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
Propylene, ≥99%
Sigma-Aldrich
4-Ethylguaiacol, ≥98%, FCC, FG
Sigma-Aldrich
1,1′-Azobis(cyclohexanecarbonitrile), 98%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
4-Ethylguaiacol, natural, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%