Merck
  • Nylon nanoweb with TiO2 nanoparticles as a solid matrix for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
Nylon nanoweb with TiO2 nanoparticles as a solid matrix for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
Rapid communications in mass spectrometry : RCM (2014-10-11)
Jo-Il Kim, Su-Yeol Ryu, Jong-Min Park, Joo-Yoon Noh, Min-Jung Kang, Seung-Yeop Kwak, Jae-Chul Pyun
ABSTRACT
The solid matrices used for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS) enable the analysis of small molecules by preventing fragmentations of organic matrix molecules in the low mass-to-charge ratio (m/z) range. In this work, a nylon nanoweb with TiO(2) particles was developed as a solid matrix for MALDI-TOFMS to improve the low intensities of mass peaks, narrow detection ranges and low signal-to-noise levels. The nylon nanoweb with TiO(2) particles was prepared by simultaneously electrospinning a nylon nanoweb and electrospraying TiO(2) nanoparticles measuring 25 nm in diameter to form TiO(2) spheres 300 nm in diameter. MS of multiple analytes was demonstrated in the low molecular weight range using eight amino acids. Additionally, leucine-enkephalin (555.6 g/mol) and cyclic citrullinated peptide (1668 g/mol) were used as model analytes to test the feasibility of a nylon nanoweb containing TiO(2) particles as a solid matrix for MALDI-TOFMS. The nylon nanoweb with TiO(2) particles can be applied for the detection of volatile small molecule analytes in the m/z ratio range of small molecules.
Product Description
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Leucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Arginine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%