Skip to Content
Merck
All Photos(3)

Key Documents

S5386

Sigma-Aldrich

DL-Serine

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)

Synonym(s):

(±)-2-Amino-3-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$49.90
100 G
$135.00

$49.90


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$49.90
100 G
$135.00

About This Item

Linear Formula:
HOCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721405
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75

$49.90


Available to ship on04 April 2025Details


Request a Bulk Order

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white

mp

240 °C (dec.) (lit.)

SMILES string

NC(CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in insect and mammalian cell culture applications where the pure L-form is not required.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Picard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 118(9), 1161-1171 (2015-03-15)
The diaphragm is a unique skeletal muscle designed to be rhythmically active throughout life, such that its sustained inactivation by the medical intervention of mechanical ventilation (MV) represents an unanticipated physiological state in evolutionary terms. Within a short period after
K U Sales et al.
Oncogene, 34(3), 346-356 (2014-01-29)
The membrane-anchored serine protease, matriptase, is consistently dysregulated in a range of human carcinomas, and high matriptase activity correlates with poor prognosis. Furthermore, matriptase is unique among tumor-associated proteases in that epithelial stem cell expression of the protease suffices to
Jacob T Bjerrum et al.
Inflammatory bowel diseases, 20(12), 2340-2352 (2014-10-31)
It is unknown why patients with extensive ulcerative colitis (UC) have a higher risk of colorectal cancer compared with patients with left-sided UC. This study characterizes the inflammatory processes in left-sided UC, pancolitis, and UC-associated dysplasia at the transcriptional level
Marne C Hagemeijer et al.
Virology, 458-459, 125-135 (2014-06-15)
Coronaviruses replicate their genomes in association with rearranged cellular membranes. The coronavirus nonstructural integral membrane proteins (nsps) 3, 4 and 6, are key players in the formation of the rearranged membranes. Previously, we demonstrated that nsp3 and nsp4 interact and
Wei Wu et al.
Molecular cancer research : MCR, 12(12), 1717-1728 (2014-07-27)
Colorectal cancer metastasis is a major cause of mortality worldwide, which may only be controlled with novel methods limiting tumor dissemination and chemoresistance. High stathmin-1 (STMN1) expression was previously established as a hallmark of colorectal cancer progression and predictor of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service