Skip to Content
Merck
All Photos(4)

Key Documents

50046

Sigma-Aldrich

Glycine

≥99.0% (NT), BioUltra

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$49.90
250 G
$172.00
1 KG
$509.00

$49.90


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$49.90
250 G
$172.00
1 KG
$509.00

About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

$49.90


Available to ship on14 April 2025Details


Request a Bulk Order

Product Name

Glycine, BioUltra, ≥99.0% (NT)

grade

Molecular Biology

Quality Level

product line

BioUltra

Assay

≥99.0% (NT)

form

crystalline

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.05% (as SO4)

loss

≤0.05% loss on drying, HV, 20-26 °C

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

1 M HCl: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NCC(O)=O

λ

1 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Glycine has been used as a component of the viral extraction mixture for adenovirus viral extraction.[1]

Biochem/physiol Actions

Glycine is a non-essential amino acid. It is involved in the synthesis of purines, serine and porphyrins. Glycine-gated channels mediate the influx of calcium through cell membranes. It also acts as a competitive agonist for glutamate in binding to the NMDA (N-methyl-D-aspartate) receptors. In cancer cells, the synthesis of glycine might be increased due to increased demand for the amino acid.[2][3]
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Cobianchi et al.
Methods in Enzymology, 152, 101-101 (1987)
Determining pathogen and indicator levels in class B municipal organic residuals used for land application.
Rhodes ER
Journal of Environmental Quality, 44(1), 265-274 (2015)
Characterization of in vitro translation products.
R Jagus
Methods in enzymology, 152, 296-304 (1987-01-01)
Metabolite Profiling Identifies a Key Role for Glycine in Rapid Cancer Cell Proliferation
Mohit Jain
Science, 336(6084), 1040-1044 (2012)
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service