Merck
  • Determination of bisphenol A, triclosan and their metabolites in human urine using isotope-dilution liquid chromatography-tandem mass spectrometry.
Determination of bisphenol A, triclosan and their metabolites in human urine using isotope-dilution liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-05-20)
Gilles Provencher, René Bérubé, Pierre Dumas, Jean-François Bienvenu, Eric Gaudreau, Patrick Bélanger, Pierre Ayotte
ABSTRACT
Bisphenol A (BPA) and triclosan (TCS) are ubiquitous environmental phenols exhibiting endocrine disrupting activities that may be involved in various health disorders in humans. There is a need to measure separately free forms and conjugated metabolites because only the former are biologically active. We have developed sensitive methods using isotope-dilution liquid chromatography-tandem mass spectrometry for individual measurements of free BPA and TCS as well as their metabolites, BPA glucuronide (BPAG), BPA monosulfate (BPAS), BPA disulfate (BPADS), TCS glucuronide (TCSG) and TCS sulfate (TCSS) in urine. Comparative analyses of urine samples from 46 volunteers living in the Quebec City area using the new methods and a GC-MS/MS method previously used in our laboratory revealed very strong correlations for total BPA (Spearman's rs=0.862, p<0.0001) and total TCS concentrations (rs=0.942, p<0.0001). Glucuronide metabolites were the most abundant BPA and TCS species in urine samples (>94% of total urinary concentrations). Unconjugated TCS concentrations represented a small proportion of total TCS species (median=1.6%) but its concentration was likely underestimated due to losses by adsorption to the surface of polypropylene tubes used for sample storage. To our knowledge, we are the first to report levels of free, sulfated and glucuronidated TCS levels in human urine.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%