Merck
  • Circulating biomarkers of one-carbon metabolism in relation to renal cell carcinoma incidence and survival.
Circulating biomarkers of one-carbon metabolism in relation to renal cell carcinoma incidence and survival.
Journal of the National Cancer Institute (2014-11-08)
Mattias Johansson, Anouar Fanidi, David C Muller, Julie K Bassett, Øivind Midttun, Stein Emil Vollset, Ruth C Travis, Domenico Palli, Amalia Mattiello, Sabina Sieri, Antonia Trichopoulou, Pagona Lagiou, Dimitrios Trichopoulos, Börje Ljungberg, Göran Hallmans, Elisabete Weiderpass, Guri Skeie, Carlos A González, Miren Dorronsoro, Petra H Peeters, H Bas Bueno-de-Mesquita, Martine M Ros, Marie-Christine Boutron Ruault, Guy Fagherazzi, Françoise Clavel, María-José Sánchez, Aurelio Barricarte Gurrea, Carmen Navarro, J Ramon Quiros, Kim Overvad, Anne Tjønneland, Krassimira Aleksandrova, Paolo Vineis, Marc J Gunter, Rudolf Kaaks, Graham Giles, Caroline Relton, Elio Riboli, Heiner Boeing, Per Magne Ueland, Gianluca Severi, Paul Brennan
ABSTRACT
The etiology of renal cell carcinoma (RCC) is only partially understood, but a metabolic component appears likely. We investigated biomarkers of one-carbon metabolism and RCC onset and survival. The European Prospective Investigation into Cancer and Nutrition (EPIC) recruited 385747 participants with blood samples between 1992 and 2000, and this analysis included 556 RCC case-control pairs. A subsequent replication study included 144 case-control pairs nested within the Melbourne Collaborative Cohort Study (MCCS). Plasma concentrations of vitamin B2, vitamin B6, folate, vitamin B12, methionine and homocysteine were measured in prediagnostic samples and evaluated with respect to RCC risk using conditional and unconditional logistic regression models, and to all-cause mortality in RCC cases using Cox regression models. All statistical tests were two-sided. EPIC participants with higher plasma concentrations of vitamin B6 had lower risk of RCC, the odds ratio comparing the 4(th) and 1(st) quartiles (OR4vs1) being 0.40 95% confidence interval [CI] = 0.28 to 0.57, P trend < .001. We found similar results after adjusting for potential confounders (adjusted P trend < .001). In survival analysis, the hazard ratio for all-cause mortality in RCC cases when comparing the 4(th) and 1(st) quartiles (HR4vs1) of vitamin B6 was 0.57 (95% CI = 0.37 to 0.87, P trend < .001). Subsequent replication of these associations within the MCCS yielded very similar results for both RCC risk (OR4vs1 = 0.47, 95% CI = 0.23 to 0.99, P trend = .07) and all-cause mortality (HR4vs1 = 0.56, 95% CI = 0.27 to 1.17, P trend = .02). No association was evident for the other measured biomarkers. Study participants with higher circulating concentrations of vitamin B6 had lower risk of RCC and improved survival following diagnosis in two independent cohorts.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard