Skip to Content
Merck
All Photos(1)

Documents

53663

Sigma-Aldrich

Activated Charcoal Norit®

greener alternative

Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

biological source

peat

grade

corresponds U.S. Food chemicals codex (3rd Ed.)

form

powder

quality

highly purified
steam activated and acid washed

manufacturer/tradename

Norit® SX ultra

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

ign. residue

≤8%

loss

≤10% loss on drying

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Charcoal Norit® (Norit SX ultra) can be used as an adsorbent for the uptake of 5-hydroxymethylfurfural (HMF) from fructose dehydration media. It can also be used in the preparation of Pd/C, Re/C and PdRe/C catalysts.

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dehydration of fructose into furans over zeolite catalyst using carbon black as adsorbent
Dornath P and Fan W
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 191, 10-17 (2014)
Vapor-phase hydrodeoxygenation of guaiacol over carbon-supported Pd, Re and PdRe catalysts
Thompson ST and Lamb HH
Applied Catalysis A: General, 563, 105-117 (2018)
Synthesis of sugar alcohols by hydrolytic hydrogenation of cellulose over supported metal catalysts.
Kobayashi H, et al.
Green Chemistry, 13(2), 326-333 (2011)
Performance of carbon nanomaterial (nanotubes and nanofibres) supported platinum and palladium catalysts for the hydrogenation of cinnamaldehyde and of 1-octyne.
Jung A, et al.
Applied Catalysis A: General, 362(1), 95-105 (2009)
Use of activated charcoal for the removal of patulin from cider.
Sands DC, et al.
Applied and Environmental Microbiology, 32(3), 388-391 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service