Skip to Content
Merck
All Photos(2)

Key Documents

242233

Sigma-Aldrich

Activated charcoal

greener alternative

DARCO®, 4-12 mesh particle size, granular

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$70.40
1 KG
$168.00

$70.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$70.40
1 KG
$168.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

$70.40


Check Cart for Availability

Request a Bulk Order

vapor pressure

<0.1 mmHg ( 20 °C)

form

granular

autoignition temp.

842 °F

manufacturer/tradename

DARCO®

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

impurities

≤0.40% dust
≤12% moisture content

particle size

4-12 mesh

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated charcoal (DARCO®, 4-12 mesh particle size, granular) can be used in combination with ozone for the tertiary treatment of wastewater. It can also be utilized in selective adsorption of phenanthrene with a selectivity of ~38.[1][2]

Other Notes

Acid-washed lignite carbon for general uses.

Legal Information

DARCO is a registered trademark of Cabot Norit Americas, Inc.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Granular activated carbon promoted ozonation of a food-processing secondary effluent
Alvarez PM, et al.
Journal of Hazardous Materials, 185(2-3), 776-783 (2011)
Single and competitive adsorption of Cd and Zn onto a granular activated carbon.
Gabaldon, et al.
Water Research, 30(12), 3050-3060 (1996)
Selective adsorption of phenanthrene dissolved in surfactant solution using activated carbon
Ahn CK, et al.
Chemosphere, 69(11), 1681-1688 (2007)
The effects of the surface oxidation of activated carbon, the solution pH and the temperature on adsorption of ibuprofen
Guedidi H, et al.
Carbon, 54, 432-443 (2013)
Yuka Kawashita et al.
Organic letters, 5(20), 3713-3715 (2003-09-26)
[reaction: see text] 2-Arylbenzoxazoles were directly synthesized from substituted 2-aminophenols and aldehydes in the presence of activated carbon (Darco KB) in xylene under an oxygen atmosphere.

Questions

1–8 of 8 Questions  
  1. what is the raw material of this product? (activated charcoal)(242233) wood, organic material

    1 answer
    1. This product is acid-washed lignite carbon (coal).

      Helpful?

  2. What is the surface area of this activated carbon?

    1 answer
    1. The surface area of this product is 600-675 m²/g.

      Helpful?

  3. I would like to know the surface area of this product ?

    1 answer
    1. The surface area of this product is 600-675 m2/g.

      Helpful?

  4. What is the typical pH of Product 242233, DARCO® Acid Washed Activated charcoal?

    1 answer
    1. The typical pH of a water extract of Product 242233, DARCO® Acid Washed Activated charcoal, is 4.6.

      Helpful?

  5. What is the purpose of the acid washing performed on Product 242233, DARCO® Acid Washed Activated Charcoal?

    1 answer
    1. The acid washing performed on Product 242233, DARCO® Acid Washed Activated Charcoal, removes minute impurities from the surface of the particles and from the pores.

      Helpful?

  6. What is the typical apparent density of product 242233, DARCO® Acid Washed Activated Charcoal?

    1 answer
    1. The typical apparent density of product 242233, DARCO® Acid Washed Activated Charcoal, is 0.4 g/ml, or 25 lb/ft3.

      Helpful?

  7. What is the typical pore volume of product 242233, DARCO® Acid Washed Activated Charcoal?

    1 answer
    1. The typical pore volume of product 242233, DARCO® Acid Washed Activated Charcoal, is 0.90 ml/g.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service