Merck
  • Ag and IL-2 immune complexes efficiently expand Ag-specific Treg cells that migrate in response to chemokines and reduce localized immune responses.
Ag and IL-2 immune complexes efficiently expand Ag-specific Treg cells that migrate in response to chemokines and reduce localized immune responses.
European journal of immunology (2013-12-18)
Ryoko Hamano, Tomohisa Baba, Soichiro Sasaki, Utano Tomaru, Akihiro Ishizu, Mitsuhiro Kawano, Masakazu Yamagishi, Naofumi Mukaida
ABSTRACT
An intravenous administration of a high-dose antigen (Ag) can induce immune tolerance and suppress the immune response, but the mechanism remains unclear. We recently proved that a combined i.v. administration of OVA and IL-2-anti-IL-2 Ab immune complexes (IL-2 ICs) efficiently expands OVA-specific Treg cells in the thymus and induces their migration into peripheral blood, by using OVA-specific TCR Tg-expressing DO11.10 mice. Here, we demonstrate that the expanded OVA-specific Treg cells rapidly move into the air pouch after OVA injection in DO11.10 mice. The migration was inhibited by blocking the axis of a chemokine receptor, CCR2. Moreover, prior treatment with OVA and IL-2 ICs enhanced OVA-specific Treg-cell migration and inhibited OVA-induced delayed-type hypersensitivity (DTH) reactions in the skin of BM chimeric mice with 15% of T cells expressing OVA-specific TCR. Blocking the CCR2 axis reversed this suppression of DTH in these mice. Furthermore, prior treatment with OVA and IL-2 ICs effectively reduced DTH reactions even in WT mice possessing only a very small population of OVA-specific T cells. Thus, the treatment with Ag and IL-2 ICs can efficiently expand Ag-specific Treg cells with the capacity to migrate and reduce localized immune responses.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)