Merck
  • Cutting edge: The signals for the generation of T cell memory are qualitatively different depending on TCR ligand strength.
Cutting edge: The signals for the generation of T cell memory are qualitatively different depending on TCR ligand strength.
Journal of immunology (Baltimore, Md. : 1950) (2013-11-19)
Karin M Knudson, Sara E Hamilton, Mark A Daniels, Stephen C Jameson, Emma Teixeiro
ABSTRACT
CD8 T cell memory critically contributes to long-term immunity. Both low- and high-affinity TCR signals are able to support the differentiation of memory CD8 T cells. However, it is unclear whether the requirements for memory development change when TCR signal strength is altered. To gain further insight into this question, we used a TCRβ transmembrane domain mutant model that is defective in the generation of memory in response to high-affinity ligands. Surprisingly, lowering TCR signal strength, by stimulation with low-affinity ligands, resulted in normal memory development. Restoration of memory correlated with recovery of TCR-dependent NF-κB signaling. Thus, these data provide novel evidence that the requirements for memory are qualitatively different depending on TCR signal strength.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)