Skip to Content
Merck
All Photos(2)

Key Documents

V900518

Sigma-Aldrich

Polysucrose 400

Vetec, reagent grade

Synonym(s):

Sucrose-epichlorohydrin copolymers

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
Product V900518 is not currently sold in your country. Contact Technical Service

grade

reagent grade

product line

Vetec

form

powder

Looking for similar products? Visit Product Comparison Guide

Application

Used for cell separation and organelle isolation.

Other Notes

A nonionic synthetic polymer of sucrose.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service