V900386
Resveratrol
Vetec™, reagent grade, 98%
Synonym(s):
3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
About This Item
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
98%
form
powder
solubility
acetone: 50 mg/mL
storage temp.
−20°C
SMILES string
Oc1ccc(cc1)\C=C\c2cc(O)cc(O)c2
InChI
1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+
InChI key
LUKBXSAWLPMMSZ-OWOJBTEDSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>392.0 °F - closed cup - (External MSDS)
Flash Point(C)
> 200.0 °C - closed cup - (External MSDS)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service