Skip to Content
Merck
Resveratrol Vetec™, reagent grade, 98%

Resveratrol

Resveratrol Vetec™, reagent grade, 98%
Synonym(s):
3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol, Resveratrol
Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Tissue
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique