Skip to Content
Merck
All Photos(2)

Key Documents

V900342

Sigma-Aldrich

Cysteamine hydrochloride

Vetec, powder or crystals, 98%

Synonym(s):

β-Mercapto­ethylamine hydrochloride, 2-Amino­ethane­thiol hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
113.61
Beilstein:
3590083
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Product V900342 is not currently sold in your country. Contact Technical Service

Product Name

Cysteamine hydrochloride, Vetec, reagent grade, 98%

grade

reagent grade

product line

Vetec

Assay

98%

form

powder or crystals

mp

67-71 °C

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].NCCS

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Zhang et al.
The Analyst, 137(20), 4822-4828 (2012-09-04)
Sol-gel copolymer-templated mesoporous silica films with a thickness of 70 nm and interpore spacing of 4.34 nm were fabricated on gold layer covered glass substrates for application as a wavelength-interrogated surface plasmon resonance (SPR) sensor. The resonance wavelength (λ(R)) of
Rajib Ghosh et al.
Biomaterials, 34(3), 807-816 (2012-11-01)
Lack of affordable technologies for delivering microRNAs and siRNAs into cells on a large scale has hindered our efforts to rapidly parse through hundreds of dysregulated genes/microRNAs in order to identify drivers of complex diseases. The instability and polyanionic nature
Bin Liu et al.
Science (New York, N.Y.), 337(6092), 351-354 (2012-07-24)
Defective catabolite export from lysosomes results in lysosomal storage diseases in humans. Mutations in the cystine transporter gene CTNS cause cystinosis, but other lysosomal amino acid transporters are poorly characterized at the molecular level. Here, we identified the Caenorhabditis elegans
Yan Yang et al.
Pharmacological research, 95-96, 71-81 (2015-04-04)
Curcumin (CUR) has been proven to be clinically effective in rheumatoid arthritis (RA) therapy, but its low oral bioavailability eclipses existent evidence that attempts to explain the underlying mechanism. Small intestine, the only organ exposed to a relatively high concentration
Adrien Jézégou et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), E3434-E3443 (2012-11-22)
Cystinosin, the lysosomal cystine exporter defective in cystinosis, is the founding member of a family of heptahelical membrane proteins related to bacteriorhodopsin and characterized by a duplicated motif termed the PQ loop. PQ-loop proteins are more frequent in eukaryotes than

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service