Skip to Content
Merck
All Photos(1)

Key Documents

30078

Sigma-Aldrich

Cysteamine hydrochloride

BioXtra

Synonym(s):

β-Mercapto­ethylamine hydrochloride, 2-Amino­ethane­thiol hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$175.00
100 G
$686.00

$175.00


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$175.00
100 G
$686.00

About This Item

Linear Formula:
HSCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
113.61
Beilstein:
3590083
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

$175.00


Available to ship on08 April 2025Details


Request a Bulk Order

product line

BioXtra

Quality Level

pH

3.5-5.0 (25 °C, 1 M in H2O)

mp

67-71 °C

solubility

H2O: 1 m at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 270 nm in H2O at 1 M

λ

1 M in H2O

UV absorption

λ: 280 nm Amax: ≤0.3

storage temp.

2-8°C

SMILES string

Cl[H].NCCS

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cysteamine is an aminothiol that can reduce important oxidized disulfide molecules such as cystine to yield cysteine.

Application

Cysteamine is used in a wide range of applications such as regulation of gene expression, depletion of somatostatin and coating of nanoparticles. It may be used to study the potential value of cystine in metabolism disorders such as cystinosis. Cysteamine may be used as a redox sensitive component in biosensor development.

Other Notes

Aminothiol radioprotector;[1] Inducer of duodenal ulcers in rodents;[2] Depletes somatostatin concentration in rat tissues.[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Widmann et al.
Journal of neurochemistry, 50(6), 1682-1686 (1988-06-01)
Cysteamine and its dimeric form cystamine have been applied to the rat striatum by local injection. Both compounds resulted in a dose-dependent decrease of somatostatin levels. Maximal reduction of somatostatin (by about 50%) was obtained at a dose of 50
D. Vasilescu et al.
International Journal of Quantum Chemistry, 14, 149-149 (1987)
Mengxiao Yu et al.
Journal of the American Chemical Society, 133(29), 11014-11017 (2011-07-01)
pH regulates many cellular processes and is also an indicator of disease progression. Therefore, pH-responsive materials often serve as either tools in the fundamental understanding of cell biology or medicine for disease diagnosis and therapy. While gold nanoparticles have broad
S Szabo et al.
Endocrinology, 109(6), 2255-2257 (1981-12-01)
Administration of cysteamine (mercaptoethylamine) induces in rats severe perforating duodenal ulcers. Because the ulcerogenic properties of cysteamine are markedly reduced by treatment with somatostatin, we considered the possibility that cysteamine-induced duodenal ulcer might be mediated by depletion of tissue somatostatin
Jamin B Hein et al.
Bio-protocol, 10(13), e3669-e3669 (2021-03-05)
Every living cell relies on signal transduction pathways comprised of protein-protein interactions (PPIs). In many cases, these PPIs are between a folded protein domain and a short linear motif (SLiM) within an unstructured region of a protein. As a result

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service