Skip to Content
Merck
All Photos(3)

Key Documents

V900334

Sigma-Aldrich

L-Glutamic acid monosodium salt hydrate

Vetec, reagent grade, ≥99%

Synonym(s):

(S)-2-Aminopentanedioic acid, L-Glutamic acid monosodium salt, Glu, MSG, Monosodium glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
169.11 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

InChI

1S/C5H9NO4.Na.H2O.H/c6-3(5(9)10)1-2-4(7)8;;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2;

InChI key

JYDGDQPVBSBYRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roie Tzadok et al.
Mycoses, 58(12), 694-698 (2015-10-03)
Invasive fungal infections are an important cause of morbidity and mortality after allogeneic haematopoietic stem cell transplantation. We evaluated, in our allogeneic stem cell transplant patients, the effect on the incidence of invasive fungal infection during neutropenia of a strategy
Signorá Peres Konrad et al.
Clinics (Sao Paulo, Brazil), 67(10), 1209-1214 (2012-10-17)
The aim of this study was to evaluate cardiovascular autonomic function in a rodent obesity model induced by monosodium glutamate injections during the first seven days of life. The animals were assigned to control (control, n = 10) and monosodium
Katie M Di Sebastiano et al.
The British journal of nutrition, 110(12), 2165-2172 (2013-06-12)
Glutamate is linked to the glycolytic process, particularly when co-ingested with carbohydrate, but its effects on glucose metabolism are poorly characterised. The present study aimed to (1) specifically examine the effects of carbohydrate administration on circulating glutamate concentrations and (2)
Karen Ackroff et al.
Physiology & behavior, 107(2), 207-217 (2012-07-11)
Monosodium glutamate (MSG), the prototypical umami substance, is used as a flavor enhancer in many foods, but when presented alone is often only weakly attractive. Yet with experience mice will develop strong preferences for MSG solution over water. The present
Cássia Borges Lima et al.
Life sciences, 93(9-11), 388-392 (2013-07-23)
Monosodium glutamate (MSG) is a neuroexcitatory amino acid used in human food to enhance flavor. MSG can affect the morphological and electrophysiological organization of the brain. This effect is more severe during brain development. Here, we investigated the electrophysiological and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service