Skip to Content
Merck
All Photos(2)

Key Documents

V800374

Sigma-Aldrich

Sodium dichromate dihydrate

AR, ≥99.5%

Synonym(s):

Sodium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2Cr2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.5%
grade:
AR
form:
crystalline powder
Product V800374 is not currently sold in your country. Contact Technical Service

grade

AR

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

3.5 (20 °C, 100 g/L)

mp

91 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.002%

SMILES string

O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1

InChI key

JYDRNIYTFCBIFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chad M Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 123(1), 58-70 (2011-06-30)
Chronic ingestion of high concentrations of hexavalent chromium [Cr(VI)] in drinking water induces intestinal tumors in mice. To investigate the mode of action (MOA) underlying these tumors, a 90-day drinking water study was conducted using similar exposure conditions as in
Silvio De Flora et al.
Mutation research, 610(1-2), 38-47 (2006-07-29)
Chromium(VI) compounds are genotoxic in a variety of cellular systems. Their potential carcinogenicity is affected by toxicokinetic patterns restricting bioavailability to certain targets, and by metabolic pathways affecting interaction of chromate-derived reactive species with DNA. Epidemiological data indicate that chromium(VI)
Chad M Thompson et al.
Regulatory toxicology and pharmacology : RTP, 64(1), 68-76 (2012-06-19)
In vitro studies on hexavalent chromium [Cr(VI)] indicate that reduced forms of this metal can interact with DNA and cause mutations. Recently, Cr(VI) was shown to induce intestinal tumors in mice; however, Cr(VI) elicited redox changes, cytotoxicity and hyperplasia -
Isabelle Hininger et al.
Free radical biology & medicine, 42(12), 1759-1765 (2007-05-22)
Several studies have demonstrated beneficial effects of supplemental trivalent Cr in subjects with reduced insulin sensitivity with no documented signs of toxicity. However, recent studies have questioned the safety of supplemental trivalent Cr complexes. The objective of this study was
Amanda J Lee et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 9(2), 103-115 (2004-09-17)
Hexavalent chromium (Cr[VI]) is a genotoxic carcinogen that has been associated with an increased risk of nasal and respiratory tract cancers following occupational exposure. Although the precise mechanism(s) remain to be elucidated, there is evidence for a role of oxidative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service