Skip to Content
Merck
All Photos(1)

Key Documents

1.06336

Supelco

Sodium dichromate dihydrate

for analysis EMSURE® ACS

Synonym(s):

Sodium dichromate dihydrate, Sodium bichromate, Sodium pyrochromate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$137.00
1 KG
$423.00

$137.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 G
$137.00
1 KG
$423.00

About This Item

Empirical Formula (Hill Notation):
Cr2Na2O7 · 2H2O
CAS Number:
Molecular Weight:
298.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-190-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid

$137.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.5-100.5% (iodometric)

form

solid

potency

50 mg/kg LD50, oral (Rat)
1000 mg/kg LD50, skin (Rabbit)

impurities

≤0.005% Insoluble matter

pH

3.5 (20 °C, 100 g/L in H2O)

bp

400 °C/1013 hPa (decomposition)

mp

356.7 °C (anhydrous substance)

density

2.35 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Al: ≤0.002%
Ca: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.002%
K: ≤0.01%
Mg: ≤0.005%
Pb: ≤0.002%

storage temp.

2-30°C

SMILES string

[Cr](=O)(=O)([O-])[O-].[Cr](=O)(=O)=O.[Na+].[Na+].O.O

InChI

1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1

InChI key

JYDRNIYTFCBIFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (iodometric): 99.5 - 100.5 %
Identity: passes test
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.01 %
Al (Aluminium): ≤ 0.002 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Pb (Lead): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service