Skip to Content
Merck
All Photos(1)

Key Documents

V800335

Sigma-Aldrich

Potassium phosphate dibasic

AR, anhydrous, ≥99%

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
anhydrous
form:
crystalline powder

grade

AR
anhydrous

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

8.7-9.3 (25 °C, 50 g/L)

anion traces

chloride (Cl-): ≤0.003%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.0005%

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Ye et al.
Biosensors & bioelectronics, 73, 208-213 (2015-06-17)
Several volatile organic compounds (VOCs) are released from human breath or skin. Like chemical substances in blood or urine, some of these vapors can provide valuable information regarding the state of the human body. A highly sensitive acetone biochemical gas
Ragnar Thomsen et al.
Drug testing and analysis, 7(7), 565-576 (2014-10-28)
Synthetic cannabimimetic agents are a large group of diverse compounds which act as agonists at cannabinoid receptors. Since 2004, synthetic cannabinoids have been used recreationally, although several of the compounds have been shown to cause severe toxicity in humans. In
Pamela Gamba et al.
PLoS genetics, 11(6), e1005047-e1005047 (2015-06-26)
Gene expression can be highly heterogeneous in isogenic cell populations. An extreme type of heterogeneity is the so-called bistable or bimodal expression, whereby a cell can differentiate into two alternative expression states. Stochastic fluctuations of protein levels, also referred to
Kenshi Hayakawa et al.
Journal of bioscience and bioengineering, 120(5), 532-538 (2015-04-29)
S-Adenosyl-L-methionine (SAM) is a major biological methyl group donor, and is used as a nutritional supplement and prescription drug. Yeast is used for the industrial production of SAM owing to its high intracellular SAM concentrations. To determine the regulation mechanisms
Audrey Beaussart et al.
ACS nano, 8(10), 10723-10733 (2014-10-07)
A variety of bacterial pathogens use nanoscale protein fibers called type IV pili to mediate cell adhesion, a primary step leading to infection. Currently, how these nanofibers respond to mechanical stimuli and how this response is used to control adhesion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service