Skip to Content
Merck
All Photos(1)

Documents

V001758

Supelco

Hydroxynaphthol blue

Synonym(s):

1-(2-Hydroxy-4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2,7-disulfonic acid trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N2Na3O11S3
CAS Number:
Molecular Weight:
620.47
EC Number:
MDL number:
PubChem Substance ID:

SMILES string

[Na+].[Na+].[Na+].Oc1cc(c2ccccc2c1\N=N\c3c(O)c(cc4cc(ccc34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H14N2O11S3.3Na/c23-15-9-16(35(28,29)30)13-3-1-2-4-14(13)18(15)21-22-19-12-6-5-11(34(25,26)27)7-10(12)8-17(20(19)24)36(31,32)33;;;/h1-9,23-24H,(H,25,26,27)(H,28,29,30)(H,31,32,33);;;/q;3*+1/p-3/b22-21+;;;

InChI key

AUIINJJXRXMPGT-ZRUFZDNISA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya Bing Duan et al.
Scientific reports, 5, 17278-17278 (2015-11-27)
Resistance of benzimidazole fungicides is related to the point mutation of the β-tubulin gene in Sclerotinia sclerotiorum. The point mutation at codon 198 (GAG → GCG, E198A) occurs in more than 90% of field resistant populations in China. Traditional detection
Ya-Bing Duan et al.
PloS one, 9(10), e111094-e111094 (2014-10-21)
Botrytis cinerea is a devastating plant pathogen that causes grey mould disease. In this study, we developed a visual detection method of B. cinerea based on the Bcos5 sequence using loop-mediated isothermal amplification (LAMP) with hydroxynaphthol blue dye (HNB). The
Xiude Hua et al.
Analytical chemistry, 86(16), 8441-8447 (2014-08-20)
Two immuno-loop-mediated isothermal amplification assays (iLAMP) were developed by using a phage-borne peptide that was isolated from a cyclic eight-peptide phage library. One assay was used to screen eight organophosphorus (OP) pesticides with limits of detection (LOD) between 2 and
Yabing Duan et al.
Scientific reports, 4, 7094-7094 (2014-11-19)
Resistance of Fusarium graminearum to carbendazim is caused by point mutations in the β2-tubulin gene. The point mutation at codon 167 (TTT → TAT, F167Y) occurs in more than 90% of field resistant isolates in China. To establish a suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service