Skip to Content
Merck
All Photos(2)

Key Documents

306509

Sigma-Aldrich

Reactive Orange 16

Dye content ≥70 %

Synonym(s):

Remazol Brilliant Orange 3R

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$132.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$132.00

About This Item

Empirical Formula (Hill Notation):
C20H17N3Na2O11S3
CAS Number:
Molecular Weight:
617.54
Colour Index Number:
17757
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$132.00


Check Cart for Availability

Request a Bulk Order

Assay

>70% (HPLC)

Quality Level

form

powder

composition

Dye content, ≥70%

mp

>300 °C (lit.)

λmax

388 nm
494 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CC(=O)Nc1ccc2cc(c(\N=N\c3ccc(cc3)S(=O)(=O)CCOS([O-])(=O)=O)c(O)c2c1)S([O-])(=O)=O

InChI

1S/C20H19N3O11S3.2Na/c1-12(24)21-15-3-2-13-10-18(36(28,29)30)19(20(25)17(13)11-15)23-22-14-4-6-16(7-5-14)35(26,27)9-8-34-37(31,32)33;;/h2-7,10-11,25H,8-9H2,1H3,(H,21,24)(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2/b23-22+;;

InChI key

DHHGSXPASZBLGC-VPMNAVQSSA-L

General description

Reactive Orange 16 is an anionic dye. It belongs to the class of azo dyes. It can be biodegraded by the yeast Consortium Spp.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hatice Atacag Erkurt
Biodegradation of Azo Dyes (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service