Skip to Content
Merck
All Photos(1)

Documents

1478232

USP

Oligosaccharide System Suitability Mixture C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-linked oligosaccharides from human α 1 acid-glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oligosaccharide sst mixture

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

mixture

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

The USP Oligosaccharide System Suitability Mixture C RS comprises N-linked oligosaccharides with bi-, tri-, and tetra-antennary structures, and varying degrees of sialylation with or without fucose, the mixture was released from human α1 acid-glycoprotein using PNGase F.

Application

Oligosaccharide System Suitability Mixture C USP reference standard suitable for use in specified USP compendial tests and assays.

Also used to prepare AB-labeled standard solutions for liquid chromatographic separation.

Additionally, used to separate Bi-, tri-, and tetra-antennary N-linked oligosaccharides with variable sialylation.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service